Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
B9D2
Entrez Gene ID
476451
Gene Name
B9 domain containing 2
Gene Aliases
-
Chromosome Location
Chr.1: 112621832 - 112627726 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005616543.3 XP_005616600.1 2 - 3 321 67
XM_005616544.3 XP_005616601.1 2 - 3 292 67
XM_533658.6 XP_533658.1 2 - 3 353 67

View Full Product Details