Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MRPL32
Entrez Gene ID
475869
Gene Name
mitochondrial ribosomal protein L32
Gene Aliases
-
Chromosome Location
Chr.18: 7212604 - 7218051 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_533077.6 XP_533077.1 - 351 71

View Full Product Details