Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAM83F
Entrez Gene ID
474500
Gene Name
family with sequence similarity 83 member F
Gene Aliases
-
Chromosome Location
Chr.10: 25106663 - 25140989 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_531728.5 XP_531728.3 4 - 5 1475 85

View Full Product Details