Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FUS
Entrez Gene ID
100856381
Gene Name
FUS RNA binding protein
Gene Aliases
-
Chromosome Location
Chr.6: 17059904 - 17071227 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005621238.3 XP_005621295.1 - 156 88

View Full Product Details