Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DPH1
Entrez Gene ID
480653
Gene Name
diphthamide biosynthesis 1
Gene Aliases
-
Chromosome Location
Chr.9: 46137573 - 46147829 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_014116841.2 XP_013972316.1 6 - 7 774 124

View Full Product Details