Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MAGOH
Entrez Gene ID
479562
Gene Name
mago homolog, exon junction complex subunit
Gene Aliases
-
Chromosome Location
Chr.5: 55688443 - 55699741 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_536702.6 XP_536702.1 1 - 2 213 97

View Full Product Details