Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TBCA
Entrez Gene ID
479173
Gene Name
tubulin folding cofactor A
Gene Aliases
-
Chromosome Location
Chr.3: 28972469 - 29027436 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_022417322.1 XP_022273030.1 - 291 87

View Full Product Details