Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TIMM17B
Entrez Gene ID
480908
Gene Name
translocase of inner mitochondrial membrane 17B
Gene Aliases
-
Chromosome Location
Chr.X: 42052592 - 42056601 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_022415554.1 XP_022271262.1 4 - 5 421 77
XM_538028.6 XP_538028.1 4 - 5 429 77

View Full Product Details