Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TTR
Entrez Gene ID
480167
Gene Name
transthyretin
Gene Aliases
-
Chromosome Location
Chr.7: 57940354 - 57947084 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_537290.5 XP_537290.1 1 - 2 137 74

View Full Product Details