Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TOMM7
Entrez Gene ID
482351
Gene Name
translocase of outer mitochondrial membrane 7
Gene Aliases
-
Chromosome Location
Chr.14: 36523707 - 36532180 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_539468.6 XP_539468.3 2 - 3 240 74

View Full Product Details