Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IL4I1
Entrez Gene ID
484371
Gene Name
interleukin 4 induced 1
Gene Aliases
-
Chromosome Location
Chr.1: 106636282 - 106673438 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005616282.2 XP_005616339.1 4 - 5 650 66
XM_014118753.2 XP_013974228.1 3 - 4 308 66
XM_022423581.1 XP_022279289.1 4 - 5 712 66

View Full Product Details