Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LAMTOR1
Entrez Gene ID
485211
Gene Name
late endosomal/lysosomal adaptor, MAPK and MTOR activator 1
Gene Aliases
-
Chromosome Location
Chr.21: 25981170 - 25986942 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_542329.6 XP_542329.2 4 - 5 534 61

View Full Product Details