Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARL16
Entrez Gene ID
483366
Gene Name
ADP ribosylation factor like GTPase 16
Gene Aliases
-
Chromosome Location
Chr.9: 513465 - 515777 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_022422629.1 XP_022278337.1 - 355 101
XM_540485.7 XP_540485.3 - 391 101

View Full Product Details