Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KLHL23
Entrez Gene ID
488392
Gene Name
kelch like family member 23
Gene Aliases
-
Chromosome Location
Chr.36: 14399025 - 14433980 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_022414920.1 XP_022270628.1 3 - 4 1499 133
XM_545514.6 XP_545514.2 3 - 4 1562 133

View Full Product Details