Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- EIF4EBP2
- Entrez Gene ID
-
489027
- Gene Name
- eukaryotic translation initiation factor 4E binding protein 2
- Gene Aliases
-
-
- Chromosome Location
-
Chr.4: 21339037 - 21364224 on Build CanFam3.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_546145.5 |
XP_546145.2 |
1 - 2 |
203 |
106 |
View Full Product Details