Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
POU2AF1
Entrez Gene ID
489413
Gene Name
POU class 2 homeobox associating factor 1
Gene Aliases
-
Chromosome Location
Chr.5: 21671000 - 21695757 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_546531.5 XP_546531.2 - 747 61

View Full Product Details