Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FARS2
Entrez Gene ID
488204
Gene Name
phenylalanyl-tRNA synthetase 2, mitochondrial
Gene Aliases
-
Chromosome Location
Chr.35: 5389694 - 5887911 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005640010.3 XP_005640067.1 4 - 5 826 64
XM_005640012.3 XP_005640069.1 4 - 5 826 64
XM_014110616.2 XP_013966091.1 4 - 5 826 64
XM_022414493.1 XP_022270201.1 4 - 5 826 64
XM_022414494.1 XP_022270202.1 5 - 6 853 64
XM_022414495.1 XP_022270203.1 3 - 4 739 64

View Full Product Details