Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DOK3
Entrez Gene ID
489087
Gene Name
docking protein 3
Gene Aliases
-
Chromosome Location
Chr.4: 35860006 - 35873539 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005619443.3 XP_005619500.1 2 - 3 2166 76

View Full Product Details