Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZMAT3
Entrez Gene ID
100855542
Gene Name
zinc finger matrin-type 3
Gene Aliases
-
Chromosome Location
Chr.34: 12527909 - 12575409 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003640141.3 XP_003640189.1 4 - 5 783 91
XM_022414143.1 XP_022269851.1 4 - 5 783 91
XM_022414144.1 XP_022269852.1 5 - 6 977 91
XM_022414145.1 XP_022269853.1 6 - 7 1089 91
XM_022414146.1 XP_022269854.1 4 - 5 719 91

View Full Product Details