Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SERTAD4
Entrez Gene ID
490276
Gene Name
SERTA domain containing 4
Gene Aliases
-
Chromosome Location
Chr.7: 8856298 - 8862280 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_547396.6 XP_547396.2 - 206 141

View Full Product Details