Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SCGB1C1
Entrez Gene ID
606779
Gene Name
secretoglobin family 1C member 1
Gene Aliases
-
Chromosome Location
Chr.18: 25351393 - 25352893 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_843182.5 XP_848275.1 2 - 3 375 81

View Full Product Details