Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EPHX4
Entrez Gene ID
490160
Gene Name
epoxide hydrolase 4
Gene Aliases
ABHD7
Chromosome Location
Chr.6: 56863779 - 56900312 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_014114843.2 XP_013970318.1 4 - 5 380 82

View Full Product Details