Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GAL3ST4
Entrez Gene ID
489843
Gene Name
galactose-3-O-sulfotransferase 4
Gene Aliases
-
Chromosome Location
Chr.6: 9361856 - 9367781 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_022419663.1 XP_022275371.1 - 486 120
XM_546961.6 XP_546961.2 - 463 120

View Full Product Details