Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMEM52
Entrez Gene ID
607284
Gene Name
transmembrane protein 52
Gene Aliases
-
Chromosome Location
Chr.5: 56900297 - 56906565 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_022418913.1 XP_022274621.1 - 214 112
XM_843905.5 XP_848998.1 - 214 112

View Full Product Details