Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DCUN1D2
Entrez Gene ID
476997
Gene Name
defective in cullin neddylation 1 domain containing 2
Gene Aliases
-
Chromosome Location
Chr.22: 60750547 - 60778245 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005634145.1 XP_005634202.1 4 - 5 650 127

View Full Product Details