Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLIC3
Entrez Gene ID
607042
Gene Name
chloride intracellular channel 3
Gene Aliases
-
Chromosome Location
Chr.9: 48634061 - 48637083 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_843599.5 XP_848692.1 2 - 3 1299 96

View Full Product Details