Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PXMP4
Entrez Gene ID
610008
Gene Name
peroxisomal membrane protein 4
Gene Aliases
-
Chromosome Location
Chr.24: 22916336 - 22930082 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_847386.5 XP_852479.2 3 - 4 448 68

View Full Product Details