Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RAB38
Entrez Gene ID
608181
Gene Name
RAB38, member RAS oncogene family
Gene Aliases
-
Chromosome Location
Chr.21: 11769924 - 11823546 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_845119.5 XP_850212.2 1 - 2 278 109

View Full Product Details