Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DGCR6L
Entrez Gene ID
608009
Gene Name
DiGeorge syndrome critical region gene 6 like
Gene Aliases
DGCR6
Chromosome Location
Chr.26: 29032221 - 29038556 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_844900.5 XP_849993.1 4 - 5 586 103

View Full Product Details