Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GLRX
Entrez Gene ID
609246
Gene Name
glutaredoxin
Gene Aliases
-
Chromosome Location
Chr.3: 13795609 - 13804732 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_846466.5 XP_851559.1 - 586 84

View Full Product Details