Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DAPL1
Entrez Gene ID
607792
Gene Name
death associated protein like 1
Gene Aliases
-
Chromosome Location
Chr.36: 4678544 - 4698711 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_844590.4 XP_849683.1 1 - 2 126 116

View Full Product Details