Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SERTAD3
Entrez Gene ID
612504
Gene Name
SERTA domain containing 3
Gene Aliases
-
Chromosome Location
Chr.1: 113258274 - 113264013 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_850231.5 XP_855324.2 1 - 2 148 84

View Full Product Details