Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MTHFS
Entrez Gene ID
610157
Gene Name
methenyltetrahydrofolate synthetase
Gene Aliases
-
Chromosome Location
Chr.3: 57480935 - 57514836 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005618427.3 XP_005618484.1 2 - 3 383 60
XM_005618428.3 XP_005618485.1 2 - 3 841 60
XM_022417012.1 XP_022272720.1 2 - 3 307 60

View Full Product Details