Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MS4A5
Entrez Gene ID
612568
Gene Name
membrane spanning 4-domains A5
Gene Aliases
-
Chromosome Location
Chr.21: 50699879 - 50717355 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_850300.5 XP_855393.1 3 - 4 454 89

View Full Product Details