Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CRYGN
Entrez Gene ID
611008
Gene Name
crystallin gamma N
Gene Aliases
-
Chromosome Location
Chr.16: 15416628 - 15427664 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_014119785.2 XP_013975260.1 - 161 97
XM_022403880.1 XP_022259588.1 - 167 97
XM_848615.3 XP_853708.1 - 165 97

View Full Product Details