Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- TIMM8B
- Entrez Gene ID
-
611348
- Gene Name
- translocase of inner mitochondrial membrane 8 homolog B
- Gene Aliases
-
-
- Chromosome Location
-
Chr.5: 21041139 - 21042769 on Build CanFam3.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_849006.5 |
XP_854099.3 |
- |
159 |
60 |
View Full Product Details