Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LUZP2
Entrez Gene ID
612193
Gene Name
leucine zipper protein 2
Gene Aliases
-
Chromosome Location
Chr.21: 45926954 - 46185667 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_014122801.2 XP_013978276.1 2 - 3 197 152
XM_022407942.1 XP_022263650.1 2 - 3 197 152

View Full Product Details