Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LRG1
Entrez Gene ID
611876
Gene Name
leucine rich alpha-2-glycoprotein 1
Gene Aliases
-
Chromosome Location
Chr.20: 55177777 - 55180382 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_849599.5 XP_854692.2 1 - 2 101 105

View Full Product Details