Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZDHHC18
Entrez Gene ID
478172
Gene Name
zinc finger DHHC-type palmitoyltransferase 18
Gene Aliases
-
Chromosome Location
Chr.2: 73274186 - 73297375 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_022414907.1 XP_022270615.1 7 - 8 816 62

View Full Product Details