Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TP53I13
Entrez Gene ID
611028
Gene Name
tumor protein p53 inducible protein 13
Gene Aliases
-
Chromosome Location
Chr.9: 43671184 - 43676330 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005624889.3 XP_005624946.1 5 - 6 934 100
XM_005624890.3 XP_005624947.1 5 - 6 928 100
XM_022423396.1 XP_022279104.1 5 - 6 893 100
XM_022423397.1 XP_022279105.1 3 - 4 464 100
XM_022423399.1 XP_022279107.1 3 - 4 292 100
XM_022423400.1 XP_022279108.1 3 - 4 291 100
XM_848643.4 XP_853736.1 3 - 4 293 100

View Full Product Details