Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CLEC12B
Entrez Gene ID
611402
Gene Name
C-type lectin domain family 12 member B
Gene Aliases
-
Chromosome Location
Chr.27: 35953961 - 35961562 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_022411026.1 XP_022266734.1 4 - 5 654 142
XM_849067.4 XP_854160.3 4 - 5 652 142

View Full Product Details