Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NCCRP1
Entrez Gene ID
612546
Gene Name
NCCRP1, F-box associated domain containing
Gene Aliases
-
Chromosome Location
Chr.1: 113989951 - 113993847 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_850276.4 XP_855369.1 5 - 6 711 136

View Full Product Details