Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GAR1
Entrez Gene ID
610515
Gene Name
GAR1 ribonucleoprotein
Gene Aliases
NOLA1
Chromosome Location
Chr.32: 29995710 - 30004545 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_014109976.2 XP_013965451.1 4 - 5 557 134
XM_014109977.2 XP_013965452.1 4 - 5 534 134
XM_014109978.2 XP_013965453.1 4 - 5 558 134
XM_014109979.2 XP_013965454.1 4 - 5 620 134
XM_014109980.2 XP_013965455.1 4 - 5 633 134
XM_014109981.2 XP_013965456.1 4 - 5 559 134
XM_848036.5 XP_853129.1 4 - 5 487 134

View Full Product Details