Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LUC7L2
Entrez Gene ID
475529
Gene Name
LUC7-like 2 (S. cerevisiae)
Gene Aliases
-
Chromosome Location
Chr.16: 9342792 - 9404797 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001131056.1 NP_001124528.1 - 1149 76
XM_005629474.2 XP_005629531.1 - 1128 76
XM_005629475.3 XP_005629532.1 - 895 76
XM_005629477.3 XP_005629534.1 - 1472 76
XM_005629478.3 XP_005629535.1 - 740 76
XM_014119776.2 XP_013975251.1 - 903 76
XM_014119777.2 XP_013975252.1 - 934 76
XM_022403592.1 XP_022259300.1 - 1631 76
XM_022403593.1 XP_022259301.1 - 1176 76
XM_022403594.1 XP_022259302.1 - 1540 76
XM_022403595.1 XP_022259303.1 - 1202 76
XM_022403596.1 XP_022259304.1 - 927 76
XM_022403597.1 XP_022259305.1 - 805 76
XM_022403598.1 XP_022259306.1 - 1029 76
XM_022403599.1 XP_022259307.1 - 874 76
XM_022403600.1 XP_022259308.1 - 797 76

View Full Product Details