Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NREP
Entrez Gene ID
100856291
Gene Name
neuronal regeneration related protein
Gene Aliases
-
Chromosome Location
Chr.3: 997142 - 1025215 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003638950.4 XP_003638998.1 - 128 62

View Full Product Details