Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WDR82
Entrez Gene ID
484740
Gene Name
WD repeat domain 82
Gene Aliases
WDR82P1
Chromosome Location
Chr.20: 37494633 - 37513746 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_854531.5 XP_859624.1 4 - 5 743 60

View Full Product Details