Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TADA3
Entrez Gene ID
476541
Gene Name
transcriptional adaptor 3
Gene Aliases
TADA3L
Chromosome Location
Chr.20: 8453269 - 8464921 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_533748.6 XP_533748.2 - 591 80

View Full Product Details