Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CRYAA
Entrez Gene ID
487786
Gene Name
crystallin alpha A
Gene Aliases
-
Chromosome Location
Chr.31: 37307070 - 37310262 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080898.1 NP_001074367.1 2 - 3 367 62
GenBank mRNA EF187819.1 - 2 - 3 367 62

View Full Product Details