Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
ARHGAP8
Entrez Gene ID
100533462
Gene Name
Rho GTPase activating protein 8
Gene Aliases
-
Chromosome Location
Chr.10: 21084626 - 21132640 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001204405.1 NP_001191334.1 10 - 10 1039 104

View Full Product Details