Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TMUB2
Entrez Gene ID
607070
Gene Name
transmembrane and ubiquitin like domain containing 2
Gene Aliases
-
Chromosome Location
Chr.9: 19204802 - 19209275 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005624403.3 XP_005624460.1 - 738 73
XM_005624405.3 XP_005624462.1 - 638 73
XM_014116520.2 XP_013971995.1 - 1186 73
XM_022422902.1 XP_022278610.1 - 1023 73
XM_022422903.1 XP_022278611.1 - 1279 73
XM_022422904.1 XP_022278612.1 - 1301 73
XM_022422905.1 XP_022278613.1 - 596 73
XM_843644.5 XP_848737.2 - 754 73

View Full Product Details